p,p,p-Trikresylfosfat, Sigma-Aldrich®
Leverantör: Merck
Fara
Synonyms: p,p,p-TCP, p,p,p-Tricresyl phosphate, Tris(4-methylphenyl) phosphate, Tri-p-tolyl phosphate, Tritolyl Phosphate
M-Clarity™ quality level = MQ200
8.14811.1000 8.14811.0250
8.14811.1000EA 1940 SEK
8.14811.1000 8.14811.0250
p,p,p-Trikresylfosfat, Sigma-Aldrich®
p,p,p-Trikresylfosfat
Formel: C₂₁H₂₁O₄P
Kokpunkt: 439 °C (1013 hPa)
Smältpunkt: 77 °C
Densitet: 1,247 g/cm³ (20 °C)
Flampunkt: 230 °C
Storage Temperature: Omgivande
CAS nummer: 78-32-0
EINECS: 215-548-8
UN: 2574
ADR: 6.1,II
Order Now
Specifikation testresultat
Solubility (20 °C) insoluble
Molar Mass 368.37 g/mol
Boiling Point 300 °C
Vapor Pressure 0.044 hPa (150 °C)
Flash Point 220 °C
Density 1.17 g/cm3 (20 °C)
Viscosity kinematic 70 mm2/s (25 °C)