Carboxymethyl cellulose sodium salt, low viscosity, Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC
217277-1 217277-250
217277-1EA 4800 SEK
217277-1 217277-250
Carboxymethyl cellulose sodium salt, low viscosity, Millipore®
Carboxymethyl cellulose sodium salt
Substrate for cellulase.
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Smältpunkt: 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00081472
CAS nummer: 9004-32-4
Merck Index: 14,01829
Order Now
Specifikation testresultat
Viscosity (2%, H₂O, 25 °C) 25 - 75 mPas