9-Acetylanthracene 96%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: 1-(9-Anthracenyl)acetone, 9-Anthryl methyl ketone, 1-(9-Anthracenyl)ethanone

L01278.06 L01278.14 L01278.PF
L01278.06EA 1100 SEK
L01278.06 L01278.14 L01278.PF
9-Acetylanthracene 96%
9-Acetylanthracene

Purity: 96% CAS No.: 784-04-3 Molecular Formula: C16H12O Formula Weight: 220.27 Melting Point: 72-76° Beilstein Reference: 1370056

Formel: C₁₆H₁₂O
Kokpunkt: 374 °C (1013 hPa)
Smältpunkt: 72…76 °C
Densitet: 1,26 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00001259
CAS nummer: 784-04-3
EINECS: 212-315-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR