(±)-3-Hydroxybutyric acid ≥80.0% (contains Polymolecular esterification product)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid
H0228-25G
H0228-5G
TCIAH0228-25GEA
3125
SEK
TCIAH0228-25G
TCIAH0228-5G
(±)-3-Hydroxybutyric acid ≥80.0% (contains Polymolecular esterification product)
(±)-3-Hydroxybutyric acid
Formel:
C₄H₈O₃ MW: 104,11 g/mol Kokpunkt: 269 °C (1013 hPa) Smältpunkt: 33…34 °C Densitet: 1,26 g/cm³ (20 °C) Storage Temperature: Kylskåp |
MDL Number:
MFCD00004546 CAS nummer: 300-85-6 EINECS: 206-099-9 Merck Index: 12,04861 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 80.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...