Triethyl orthoacetate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1,1,1-Triethoxyethane
L06004.AE L06004.AP L06004.0F
L06004.AEEA 745 SEK
L06004.AE L06004.AP L06004.0F
Triethyl orthoacetate ≥98%
Triethyl orthoacetate
Purity: 98% CAS No.: 78-39-7 Molecular Formula: CH3C(OCH2CH3)3 Formula Weight: 162.23 Flash Point: 32°(89°F) Refractive Index: 1.3970 Boiling Point: 142-143° Beilstein Reference: 506201 UN No.: 1993 MDL No.: MFCD00009223
Formel: C₈H₁₈O₃
MW: 162,23 g/mol
Kokpunkt: 142…143 °C
Densitet: 0,887
Flampunkt: 32 °C (89 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009223
CAS nummer: 78-39-7
EINECS: 201-112-4
ADR: 3,III