Metanol, torka, Hydranal™ medium for volumetric one-component Karl Fischer titration, Fluka™
Leverantör: Honeywell Chemicals
Fara
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
34741-1L 34741-2.5L 34741-6X1L 34741-4X2.5L
34741-1LEA 310 SEK
34741-1L 34741-2.5L 34741-6X1L 34741-4X2.5L
Metanol, torka, Hydranal™ medium for volumetric one-component Karl Fischer titration, Fluka™
Metanol
Suitability: use tested, Reagent for volumetric one-component KF titration (working medium)
Formel: CH₃OH
MW: 32,04 g/mol
Kokpunkt: 64-65 °C (1,013 hPa)
Smältpunkt: -98 °C
Densitet: 0,79 g/cm³ (20 °C)
Flampunkt: 11 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Suitability (for KF-titration) test passed
Water (Karl Fischer) Max. 0.01 %
Number Of Samples Min. 2