Diethoxy(dimethyl)silane 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Dimethyldiethoxysilane
174635000 174631000
ACRO174635000EA 2925 SEK
ACRO174635000 ACRO174631000
Diethoxy(dimethyl)silane 97%
Diethoxy(dimethyl)silane
Formel: C₆H₁₆O₂Si
MW: 148.28 g/mol
Kokpunkt: 114
Smältpunkt: –70 °C
Densitet: 0.86 g/cm³
Flampunkt: 11 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009068
CAS nummer: 78-62-6
EINECS: 201-127-6
UN: 2380
ADR: 3 II
Order Now
Specifikation testresultat
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.3805 to 1.3825 (20°C, 589 nm)
GC ≥96.0 %