Coomassie® Brilliant Blue R-250
Leverantör: PanReac AppliChem
Synonyms: Serva Blue R, Brillant blue R, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1), Coomassie Blue R 250, Brilliant Cyanine 6B, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, Acid Blue 83, Brilliant Blue R250, Brilliant Blue R, Brilliant indocyanine 6B
A1092.0025 A1092.0100
A1092.0025EA 805 SEK
A1092.0025 A1092.0100
Coomassie® Brilliant Blue R-250
Coomassie® Brilliant Blue R-250
Formel: C₄₅H₄₄N₃NaO₇S₂
MW: 825,98 g/mol
Smältpunkt: 174…180 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041762
CAS nummer: 6104-59-2
EINECS: 228-060-5