Nuclear Fast Red, Sigma-Aldrich®
Leverantör: SIGMA-ALDRICH MICROSCOPY
Synonyms: CI 60760, Sodium 1-amino-2,4-dihydroxy-9,10-dihydro-9,10-dioxoanthracene-3-sulphonate, Helio Fast Rubin BBL
60700-25G 60700-5G
60700-25GEA 3830 SEK
60700-25G 60700-5G
Nuclear Fast Red, Sigma-Aldrich®
Nuclear Fast Red
Formel: C₁₄H₈NNaO₇S
MW: 357,28 g/mol
Smältpunkt: 300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00078493
CAS nummer: 6409-77-4
EINECS: 229-088-0
Merck Index: 13,06512
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caeeeaeb568fad8a74fa0d222037e34b7403d835&vid=caeeeaeb568fad8a74fa0d222037e34b7403d835&prid=web&clid=SAPDAM