1,2-Dietoxietan, Sigma-Aldrich®

Leverantör: Merck
Fara

Synonyms: Diethyl glycol, Ethylene glycol diethyl ether

M-Clarity™ quality level = MQ200

8.20390.0250
8.20390.0250EA 1150 SEK
8.20390.0250
1,2-Dietoxietan, Sigma-Aldrich®
1,2-Dietoxietan
Formel: C₆H₁₄O₂
MW: 118,18 g/mol
Kokpunkt: 121,4 °C (1013 hPa)
Smältpunkt: –74 °C
Densitet: 0,8484 g/cm³ (20 °C)
Flampunkt: 35 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009253
CAS nummer: 629-14-1
EINECS: 211-076-1
UN: 1153
ADR: 3,II

Order Now

Specifikation testresultat

Solubility 34 g/l (20 °C)
Melting Point -74 °C
Molar Mass 118.17 g/mol
Boiling Point 120 - 122 °C (1013 hPa)
Vapor Pressure 12.5 hPa (20 °C)
Flash Point 35 °C
Refractive Index 1.3922 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)
Ignition Point 208 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR