Isopropyl-β-D-thiogalactopyranoside
Leverantör: Ambion
9462 9464
AMBI9462EA 0 SEK
AMBI9462 AMBI9464
Isopropyl-β-D-thiogalactopyranoside
Isopropyl-β-D-thiogalactopyranoside
Formel: C₉H₁₈O₅S
Kokpunkt: 402 °C (1013 hPa)
Storage Temperature: Frys
MDL Number: MFCD00063273
CAS nummer: 367-93-1
EINECS: 206-703-0