Isopropyl-β-D-thiogalactopyranoside, Millipore®
Leverantör: Merck Millipore Calbiochem
Synonyms: IPTG
420322-1 420322-10 420322-25 420322-5
420322-1EA 2350 SEK
420322-1 420322-10 420322-25 420322-5
Isopropyl-β-D-thiogalactopyranoside, Millipore®
Isopropyl-β-D-thiogalactopyranoside
Formel: C₉H₁₈O₅S
Kokpunkt: 402 °C (1013 hPa)
Storage Temperature: Frys
MDL Number: MFCD00063273
CAS nummer: 367-93-1
EINECS: 206-703-0