2-Fenyl-1,3-propandiol ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Phenylpropane-1,3-diol
P1249-25G
P1249-5G
TCIAP1249-25GEA
4650
SEK
TCIAP1249-25G
TCIAP1249-5G
2-Fenyl-1,3-propandiol ≥98.0%
2-Fenyl-1,3-propandiol
Formel:
HOH₂CCH(C₆H₅)CH₂OH MW: 152,19 g/mol |
MDL Number:
MFCD00236056 CAS nummer: 1570-95-2 EINECS: 411-810-2 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 50.0 to 54.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...