Bottles, narrow neck, slope shoulder, round, LDPE, with screw caps
Leverantör: PLASTEX
4320 645
215-2479EA 5100 SEK
215-2479 215-2483
Bottles, narrow neck, slope shoulder, round, LDPE, with screw caps
Flaskor Flaskor, plast
LDPE.
  • Flexible
  • Good chemical and temperature resistance
  • Slope shoulder for smooth pouring
Bottles have sloping shoulders reducing the tendency for residue deposits and tagging points on the shoulders and caps to ensure tamper proof closure.