BD Eclipse™ Needles
Leverantör: BD Medical
305781 305892 305770 305771 305895 305784 305760 305891 305894
BDAM305781EA 535 SEK
BDAM305781 BDAM305892 BDAM305770 BDAM305771 BDAM305895 BDAM305784 BDAM305760 BDAM305891 BDAM305894
BD Eclipse™ Needles
Kanyler
The BD Eclipse™ SmartSlip™ is a single use sterile needle with a safety mechanism that covers the needle point after use. In the activated position, the needle cover guards against accidental needlestick during normal handling and disposal of the used needle/syringe.
  • The SmartSlip™ Technology is specially designed for safe assembling of BD Eclipse™ needles to Luer slip syringes
  • Bevel oriented to the safety cover to allow low angle injections
  • By assembling BD Eclipse™ needles to a luer slip syringe, an audible click indicates a safely assembled needle
  • Safety can be activated immediately after injection
  • Wide, textured finger pad for thumb or forefinger
  • Single-handed with one finger activation of the safety shield, audible click indicates that the cover has safely engaged the needle
  • Natural Rubber Latex is not part of the material formulation
  • Sterile, single use
The SmartSlip™ Technology consists of an adaptor to enable connection of the BD Eclipse safety needle to a Luer slip.