cis,cis-9,12-Octadecadien-1-ol 95%
Leverantör: Thermo Fisher Scientific
Synonyms: (9Z,12Z)-Octadeca-9,12-dien-1-ol, Linoleyl Alcohol, cis,cis-9,12-Octadecadienyl-1-ol
474530050 474530250
474530050.EA 2500 SEK
474530050. 474530250.
cis,cis-9,12-Octadecadien-1-ol 95%
cis,cis-9,12-Octadecadien-1-ol
Formel: C₁₈H₃₄O
Kokpunkt: 374°C (760mmHg)
Flampunkt: 124°C
Storage Temperature: Omgivande
MDL Number: MFCD00056667
CAS nummer: 506-43-4
EINECS: 208-038-1
Order Now
Specifikation testresultat
Appearance colorless to light yellow liquid
Infrared spectrum Conforms
GC ≥94 %