Azobenzene-4,4'-dicarboxylic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Azobenzene-4,4'-dicarboxylic Acid, 4,4'-Azodibenzoic acid, 4,4'-Azodibenzoic Acid, 4,4'-(Diazene-1,2-diyl)dibenzoic acid
474480010 474480050
474480010.EA 790 SEK
474480010. 474480050.
Azobenzene-4,4'-dicarboxylic acid 98%
Azobenzene-4,4'-dicarboxylic acid
Formel: C₁₄H₁₀N₂O₄
Kokpunkt: 541°C (760mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00496621
CAS nummer: 586-91-4
EINECS: 209-589-0
Order Now
Specifikation testresultat
Appearance light yellow to brown powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %