4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride 98%
Leverantör: Thermo Fisher Scientific
Synonyms: BPADA, 4,4'-(4,4'-Isopropylidenediphenoxy)bis(phthalic anhydride), 5,5'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))bis(isobenzofuran-1,3-dione)
474470500 474472500
474470500.EA 1100 SEK
474470500. 474472500.
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride 98%
4,4'-(4,4'-Isopropylidenediphenoxy)diphthalic anhydride
Formel: C₃₁H₂₀O₈
MW: 520.49 g/mol
Kokpunkt: 712°C (760mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00319167
CAS nummer: 38103-06-9
EINECS: 253-781-7
Order Now
Specifikation testresultat
Appearance almost white to yellow powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %