1,3,5-Triiodobenzene 95%
Leverantör: Thermo Fisher Scientific
Varning
473290010 473290050
473290010.EA 2300 SEK
473290010. 473290050.
1,3,5-Triiodobenzene 95%
1,3,5-Triiodobenzene
Formel: C₆H₃I₃
MW: 455.80 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02690052
CAS nummer: 626-44-8
Order Now
Specifikation testresultat
Appearance white to almost white solid
Infrared spectrum Conforms
HPLC ≥94 %
Melting point 180°C to 186°C