Diklorättiksyra 99%, technical
Leverantör: Thermo Scientific
|
Fara
|
Synonyms:
DCA
473140010
473140025
473140010.EA
2250
SEK
473140010.
473140025.
Diklorättiksyra 99%, technical
Diklorättiksyra
Formel:
C₂H₂Cl₂O₂ MW: 128,94 g/mol Kokpunkt: 194°C (760mmHg) Densitet: 1,56 Flampunkt: >112°C Storage Temperature: Omgivande |
MDL Number:
MFCD00004223 CAS nummer: 79-43-6 EINECS: 201-207-0 UN: 1764 ADR: 8,II |
Specifikation testresultat
Appearance | clear colorless to dark yellow liquid |
Infrared spectrum | Conforms |
Titration with NaOH | 98.5 to 101.5 % |
Refractive index | 1.4650 to 1.4670 (20°C, 589 nm) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...