3,5,7,4'-Tetrahydroxy-3'-methoxyflavone 97%
Leverantör: Thermo Scientific
Synonyms: 3'-Methoxy-3,4',5,7-tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-chromen-4-one, Isorhamnetin, 3'-Methylquercetin, 3-Methylquercetin
471780100 471781000
471780100.EA 3100 SEK
471780100. 471781000.
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone 97%
3,5,7,4'-Tetrahydroxy-3'-methoxyflavone
Formel: C₁₆H₁₂O₇
MW: 316,27 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00017310
CAS nummer: 480-19-3
EINECS: 207-545-5
Order Now
Specifikation testresultat
Appearance light yellow to amber powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤0.5 %