Carprofen 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 2-(6-Chloro-9H-carbazol-2-yl)propanoic acid, 6-Chloro-alpha-methyl-9H-carbazole-2-acetic Acid
471140010 471140050
471140010.EA 915 SEK
471140010. 471140050.
Carprofen 98%
Carprofen
Formel: C₁₅H₁₂ClNO₂
MW: 273.72 g/mol
Kokpunkt: 509.1°C
Storage Temperature: Omgivande
MDL Number: MFCD00079028
CAS nummer: 53716-49-7
EINECS: 258-712-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,01876
Order Now
Specifikation testresultat
Appearance white to beige powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 195°C to 199°C
Water ≤2 %