3-Bromomethyl-3-methyloxetane 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-(Bromomethyl)-2-methyl-1,3-epoxypropane, 3-(Bromomethyl)-3-methyloxetane
470490010 470490050
470490010.EA 1275 SEK
470490010. 470490050.
3-Bromomethyl-3-methyloxetane 95%
3-Bromomethyl-3-methyloxetane
Formel: C₅H₉BrO
MW: 165.04 g/mol
Kokpunkt: 162°C (760mmHg)
Flampunkt: 62°C
Storage Temperature: Omgivande
MDL Number: MFCD02684290
CAS nummer: 78385-26-9
Order Now
Specifikation testresultat
Appearance colorless to light yellow liquid
Infrared spectrum Conforms
GC ≥94 %
Refractive index 1.4800 to 1.4840 (20°C, 589 nm)