Amlodipine maleate
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate maleate
468320010 468320050
468320010.EA 2000 SEK
468320010. 468320050.
Amlodipine maleate
Amlodipine maleate
Formel: C₂₄H₂₉ClN₂O₉
Storage Temperature: Kylskåp
MDL Number: MFCD02259457
CAS nummer: 88150-47-4
Order Now
Specifikation testresultat
Appearance white to light yellow solid
Infrared spectrum Conforms
HPLC ≥98.5 %
Melting point 160°C to 165°C