Carisoprodol
Leverantör: Thermo Scientific
Varning
Synonyms: Isopropylcarbamic acid 2-[(Carbamoyloxy)methyl]-2-methylpentyl ester, 2-[(Carbamoyloxy)methyl]-2-methylpentyl Isopropylcarbamate
466240010 466240050 466240250
466240010.EA 665 SEK
466240010. 466240050. 466240250.
Carisoprodol
Carisoprodol
Formel: C₁₂H₂₄N₂O₄
MW: 260,33 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00057661
CAS nummer: 78-44-4
EINECS: 201-118-7
Order Now
Specifikation testresultat
Appearance white to almost white powder
Infrared spectrum Conforms
Melting point 92°C to 96°C
Loss on drying ≤0.5 % (1 g, 3 hr) (vacuum)
TLC ≥97.5 %