NBQX 98+%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo(f)-quinoxaline, 6-nitro-7-sulphamoylbenzo(f)-quinoxaline-2,3-dione, 2,3-Dihydroxy-6-nitro-7-sulphamoylbenzo[f]quinoxaline-2,3-dione, NBQX, 6-Nitro-2,3-dioxo-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulphonamide
329260250 329261000
329260250.EA 7825 SEK
329260250. 329261000.
NBQX 98+%
NBQX
Formel: C₁₂H₈N₄O₆S
MW: 336,28 g/mol
Kokpunkt: 613°C
Storage Temperature: Kylskåp
MDL Number: MFCD12910445
CAS nummer: 118876-58-7
Order Now
Specifikation testresultat
Appearance almost-white to yellow powder
Infrared spectrum Conforms
HPLC ≥98 %
Water ≤5 %