Silvernitrat 99+%, meets analytical specification of Ph.Eur.
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
194050500 194052500
194050500.EA 4175 SEK
194050500. 194052500.
Silvernitrat 99+%, meets analytical specification of Ph.Eur.
Silvernitrat
Formel: AgNO₃
MW: 169.87 g/mol
Kokpunkt: 444°C (760mmHg)
Smältpunkt: 210
Storage Temperature: Omgivande
MDL Number: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1 II
Merck Index: 13,08591
Order Now
Specifikation testresultat
Appearance white to almost white transparant colorless crystalline powder or crystals
Titration with NH4SCN 99.0 to 100.5 %
Acidity/alkalinity Passes test
Appearance of solution (2 g in 50 ml water) Clear and colorless