Kaliumdikromat 106 g/L 0.36 mol/l aqueous solution Reag. Ph. Eur. 1069501
Leverantör: VWR Chemicals
Fara
87716.260EA 2070 SEK
87716.260 87716.180
Kaliumdikromat 106 g/L 0.36 mol/l aqueous solution Reag. Ph. Eur. 1069501
Kaliumdikromat
Beställningsinformation: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formel: K₂Cr₂O₇
MW: 294.18 g/mol
Kokpunkt: 500
Smältpunkt: 398
Storage Temperature: Omgivande
MDL Number: MFCD00011367
CAS nummer: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1 III
Merck Index: 12,07790