, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverantör: Avantor
Synonyms: 4-Nitrophenyl phosphate disodium salt hexahydrate, Phosphatase substrate, p-Nitrophenyl phosphate disodium salt hexahydrate, Phosphoric acid 4-nitrophenyl ester disodium salt hexahydrate, 4-Nitrophenyl phosphate disodium hexahydrate, pNPP disodium salt hexahydrate, di-Sodium 4-nitrophenyl phosphate hexahydrate
3351.0005EA 0 SEK
3351.0005 3351.0025
, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Dinatrium 4-nitrofenylfosfat hexahydrat (pNPP)
Formel: C₆H₄NNa₂O₆P·6H₂O
MW: 371.144 g/mol
Smältpunkt: 300
Storage Temperature: Frys
MDL Number: MFCD00066288
CAS nummer: 4264-83-9
EINECS: 224-246-5
Order Now
Specifikation testresultat
Assay about 70%
Free p-nitrophenol max. 0.07 %