N-Octyl-β-D-glucopyranoside
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
L13259.03 L13259.MD
L13259.03EA 2175 SEK
L13259.03 L13259.MD
N-Octyl-β-D-glucopyranoside
N-Octyl-β-D-glucopyranoside
CAS No.: 29836-26-8 Molecular Formula: C14H28O6 Formula Weight: 292.37 Melting Point: 103-104° Beilstein Reference: 84118 Optical Rotation: -31° (c=2 in methanol)
Formel: C₁₄H₂₈O₆
MW: 292.37 g/mol
Smältpunkt: 103…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00063288
CAS nummer: 29836-26-8
EINECS: 249-887-8