Formalin 10% aqueous phosphate buffer (pH 7.0 ± 0.2)
Leverantör: FF CHEMICALS
|
Varning
|
Synonyms:
Formaldehyd
FF1150035
FF1209010
FF22023000
FF22589200
FF8370150
FF8860080
FFCHFF1150035EA
0
SEK
FFCHFF1150035
FFCHFF1209010
FFCHFF22023000
FFCHFF22589200
FFCHFF8370150
FFCHFF8860080
Formalin 10% aqueous phosphate buffer (pH 7.0 ± 0.2)
Formalin
(= 4% Formaldehyde solution)
Formel:
CH₂O MW: 30.03 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00003274 CAS nummer: 50-00-0 EINECS: 200-001-8 |