Natriumdiklorisocyanurat, (max. 3% H₂O) 97%
Leverantör: Thermo Fisher Scientific
Fara
B23504.14 B23504.22 B23504.36
B23504.14EA 705 SEK
B23504.14 B23504.22 B23504.36
Natriumdiklorisocyanurat, (max. 3% H₂O) 97%
Natriumdiklorisocyanurat
Purity: 97% CAS No.: 2893-78-9 Molecular Formula: C3Cl2N3NaO3 Formula Weight: 219.95 Odour: Chlorine-like Melting Point: ca 250° dec. Beilstein Reference: 4056155 UN No.: 2465 MDL No.: MFCD00006036
Formel: C₃Cl₂N₃NaO₃
Smältpunkt: ∼ 250 °C (dec.)
Densitet: 0,96 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00006036
CAS nummer: 2893-78-9
EINECS: 220-767-7
UN: 2465
ADR: 5.1,II