Bis(trimethylsilyl)carbodiimide ≥97%
Leverantör: Thermo Scientific
Varning
A11670.06 A11670.14
A11670.06EA 2175 SEK
A11670.06 A11670.14
Bis(trimethylsilyl)carbodiimide ≥97%
Bis(trimethylsilyl)carbodiimide
Purity: 97% CAS No.: 1000-70-0 Molecular Formula: C7H18N2Si2 Formula Weight: 186.41 Flash Point: 42°(107°F) Refractive Index: 1.4340 Odour: Mild Melting Point: -23° Boiling Point: 163-164° Beilstein Reference: 2042082 UN No.: 1993 MDL No.: MFCD00051538
Formel: C₇H₁₈N₂Si₂
MW: 186,4 g/mol
Kokpunkt: 163…164 °C
Smältpunkt: –23 °C
Densitet: 0,823
Flampunkt: 42 °C (107 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00051538
CAS nummer: 1000-70-0
EINECS: 213-673-2
ADR: 3,III