X-Gal
Leverantör: Biotium
Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
10011-1 10011-2 10011
10011-1EA 2075 SEK
10011-1 10011-2 10011.
X-Gal
X-Gal
Formel: C₁₄H₁₅BrClNO₆
MW: 408,63 g/mol
Smältpunkt: 237…239 °C
Storage Temperature: Frys
MDL Number: MFCD00005666
CAS nummer: 7240-90-6
EINECS: 230-640-8