5-Amino-2-naphthalenesulphonic acid ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 5-Amino-β-naphthalenesulphonic acid, 5-Aminonaphthalene-2-sulphonic acid

A0346-500G A0346-25G
TCIAA0346-500GEA 2775 SEK
TCIAA0346-500G TCIAA0346-25G
5-Amino-2-naphthalenesulphonic acid ≥97.0%
5-Amino-2-naphthalenesulphonic acid
Formel: C₁₀H₉NO₃S
MW: 223,25 g/mol
MDL Number: MFCD00004030
CAS nummer: 119-79-9
EINECS: 204-351-2
Merck Index: 14,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31cef5894840426a82e876045fe87cbdbad38c1e&vid=31cef5894840426a82e876045fe87cbdbad38c1e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 94.0 area%
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR