4-Amino-3-hydroxinaftalen-1-sulfonsyra 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid
149271000 149270250
ACRO149271000EA 2650 SEK
ACRO149271000 ACRO149270250
4-Amino-3-hydroxinaftalen-1-sulfonsyra 95%
4-Amino-3-hydroxinaftalen-1-sulfonsyra
Formel: C₁₀H₉NO₄S
MW: 239.25 g/mol
Kokpunkt: 466
Smältpunkt: 295 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00004019
CAS nummer: 116-63-2
EINECS: 204-147-3
Merck Index: 13,00453
Order Now
Specifikation testresultat
Appearance Pink to light purple Granular powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Sulfated ash ≤0.5 %
Titration with NaOH ≥94 %
Specific optical rotation +22.5° to +24.5° (20°C, 589 nm) (c=1 in methanol)