Tetrazoliumblåklorid
Leverantör: Thermo Scientific
Synonyms: Blue Tetrazolium, 3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3'-(3,3'-Dimethoxy-4,4'-biphenylene)bis(2,5-diphenyl-2H-tetrazolium chloride), BTC, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), Blue Tetrazolium chloride
A12502.03 A12502.06 A12502.14
A12502.03EA 945 SEK
A12502.03 A12502.06 A12502.14
Tetrazoliumblåklorid
Tetrazoliumblåklorid
CAS No.: 1871-22-3 Molecular Formula: C40H32Cl2N8O2 Formula Weight: 727.66 Melting Point: ca 255° dec. Beilstein Reference: 3894077 Merck Reference: 14,9244 MDL No.: MFCD00040933
Formel: C₄₀H₃₂Cl₂N₈O₂
MW: 727,65 g/mol
Smältpunkt: ∼ 255 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00040933
CAS nummer: 1871-22-3
EINECS: 217-488-8
Merck Index: 14,9244