Nitroblå tetrazolium 90%
Leverantör: Thermo Scientific
Varning
Synonyms: 5,5'-Diphenyl-3,3'-bis(4-nitrophenyl)-2,2'-(3,3'-dimethoxybiphenyl-4,4'-ylene)ditetrazolium dichloride
415920010 415925000
ACRO415920010EA 3625 SEK
ACRO415920010 ACRO415925000
Nitroblå tetrazolium 90%
Nitroblå tetrazolium
Formel: C₄₀H₃₀Cl₂N₁₀O₆
MW: 817,65 g/mol
Smältpunkt: 189 °C
Storage Temperature: Frys
MDL Number: MFCD00012159
CAS nummer: 298-83-9
EINECS: 206-067-4
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥88 %
Titration with HClO4 ≥96.0 %