EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) Crosslinker
Leverantör: G Biosciences
Fara
Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
BC25-1 BC25-5
BC25-1EA 750 SEK
BC25-1 BC25-5
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) Crosslinker
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
EDC is a heterobifunctional, water-soluble, zero-length carbodiimide crosslinker that is used to couple carboxyl groups to primary amines. EDC activates carboxyl groups first and forms amine reactive O-acylisourea imtermediate that spontaneously reacts with primary amines to form an amide bond and isourea by-product.
The unstable nature of the intermediate in aqueous solutions makes 2-step coupling, however in conjunction with N-hydroxysuccinimide, a 2-step coupling is possible.
EDC is ideal for peptide immobilization and hapten-carrier protein conjugation
Formel: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Smältpunkt: ∼ 113 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00012503
CAS nummer: 25952-53-8
EINECS: 247-361-2
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM