Bacterial identification, Diatabs
Leverantör: ROSCO
40211
40411
40511
41811
42511
42611
42711
43711
76312
44211
44311
44611
45011
46711
46811
46911
50211
50311
50411
50511
50611
52611
52811
52911
53011
53111
53211
53311
53711
53811
53911
56211
56611
56711
56811
57511
57611
57711
57811
58411
59011
59311
59611
59811
52011
10311
10411
10511
45511
48811
52111
54011
55911
45411
57011
ROSC40211EA
3575
SEK
ROSC40211
ROSC40411
ROSC40511
ROSC41811
ROSC42511
ROSC42611
ROSC42711
ROSC43711
ROSC76312
ROSC44211
ROSC44311
ROSC44611
ROSC45011
ROSC46711
ROSC46811
ROSC46911
ROSC50211
ROSC50311
ROSC50411
ROSC50511
ROSC50611
ROSC52611
ROSC52811
ROSC52911
ROSC53011
ROSC53111
ROSC53211
ROSC53311
ROSC53711
ROSC53811
ROSC53911
ROSC56211
ROSC56611
ROSC56711
ROSC56811
ROSC57511
ROSC57611
ROSC57711
ROSC57811
ROSC58411
ROSC59011
ROSC59311
ROSC59611
ROSC59811
ROSC52011
ROSC10311
ROSC10411
ROSC10511
ROSC45511
ROSC48811
ROSC52111
ROSC54011
ROSC55911
ROSC45411
ROSC57011
Bacterial identification, Diatabs
Testsystem för mikrobiologi
Enzymatiska tester
Diatabs offers an accurate and rapid identification of clinically relevant bacteria and yeasts. The concept of individual tests - used as single tests or in combination - offers the user many advantages over traditional methods. The Diatabs system offers the opportunity of finding the most appropriate and economical tests for a number of purposes.
- Rapid (4 hour) enzymatic and modified conventional test
- Detection of natural susceptibility patterns or growth factor requirement
- An extensive range of more than 80 individual tests