Neo-Sensitabs™
Leverantör: ROSCO
79012
61312
62212
62912
74312N
74412N
76312N
77412N
62612
62812
62712
63212
63612
64012
64212
64312
60112
60212
60512
60812
61012
61712
62112
64512
70812N
74812N
75112N
75512N
62312
61112
63712
66312
75912
81812N
81912N
82012N
82212N
85112
63012
68812
98012
ROSC79012EA
84
SEK
ROSC79012
ROSC61312
ROSC62212
ROSC62912
ROSC74312N
ROSC74412N
ROSC76312N
ROSC77412N
ROSC62612
ROSC62812
ROSC62712
ROSC63212
ROSC63612
ROSC64012
ROSC64212
ROSC64312
ROSC60112
ROSC60212
ROSC60512
ROSC60812
ROSC61012
ROSC61712
ROSC62112
ROSC64512
ROSC70812N
ROSC74812N
ROSC75112N
ROSC75512N
ROSC62312
ROSC61112
ROSC63712
ROSC66312
ROSC75912
ROSC81812N
ROSC81912N
ROSC82012N
ROSC82212N
ROSC85112
ROSC63012
ROSC68812
ROSC98012
Neo-Sensitabs™
Testsystem för mikrobiologi
Antibiotikatester
Neo-Sensitabs are tablets manufactured by a process using crystalline antimicrobials. Extensive range - over 125 different antimicrobial agents and combination tablets are available. Detection of mechanisms of resistance including MRSA, ESBL, AmpC Inducers and Inducible Clindamycin resistance. Logical 5-figure alpha-numeric code, easy and safe identification of the individual tablet, optically readable by automatic reader systems. Neo-Sensitabs are standardized to CLSI recommendations and adjusted to other national guidelines.
- Long shelf life - up to five years
- Large easy-to-read printed codes
- Uniform and reliable results at all times, no loss of activity due to degradation of the antibiotics
- Store and use at room temperature without degradation of the antimicrobial agent