Lithium triethylborohydride lösning 1M i THF
Leverantör: Thermo Fisher Scientific
176991000
176998000
176991000.EA
1725
SEK
176991000.
176998000.
Lithium triethylborohydride lösning 1M i THF
Lithium triethylborohydride
Formel:
C₆H₁₆BLi Densitet: 9200 g/cm³ |
MDL Number:
MFCD00011703 CAS nummer: 22560-16-3 UN: 3148 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...