Eosin B, Sigma-Aldrich®
Leverantör: SIGMA-ALDRICH MICROSCOPY
Synonyms: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
M-Clarity™ quality level = MQ200
861006-100G 861006-10G 861006-25G
861006-100GEA 6400 SEK
861006-100G 861006-10G 861006-25G
Eosin B, Sigma-Aldrich®
Eosin B
Formel: C₂₀H₆Br₂N₂Na₂O₉
MW: 624,06 g/mol
Smältpunkt: > 310 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005041
CAS nummer: 548-24-3
EINECS: 208-943-1
Merck Index: 12,03638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b44ed31611ccd8273c2aa49e942783c769935dd&vid=7b44ed31611ccd8273c2aa49e942783c769935dd&prid=web&clid=SAPDAM