Methyl-3-hydroxy-4-nitrobenzoate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3-hydroxy-4-nitrobenzoate
A15329.06 A15329.14 A15329.22
A15329.06EA 1100 SEK
A15329.06 A15329.14 A15329.22
Methyl-3-hydroxy-4-nitrobenzoate ≥98%
Methyl-3-hydroxy-4-nitrobenzoate
Purity: 98% CAS No.: 713-52-0 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 89-92° Beilstein Reference: 2583832
Formel: C₈H₇NO₅
MW: 197,15 g/mol
Smältpunkt: 89…92 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016997
CAS nummer: 713-52-0