Butyl (±)-lactate 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Butyl 2-hydroxypropanoate, n-Butyl lactate, Butyl (±)-2-hydroxypropionate, (±)-Butyl lactate
250185000 250180025
ACRO250185000EA 1525 SEK
ACRO250185000 ACRO250180025
Butyl (±)-lactate 99%
Butyl (±)-lactate
Formel: C₇H₁₄O₃
MW: 146,19 g/mol
Kokpunkt: 185…187 °C (760 mmHg)
Smältpunkt: –43 °C
Densitet: 0,98 g/cm³
Flampunkt: 71 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004519
CAS nummer: 138-22-7
EINECS: 205-316-4
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4220 (20°C, 589 nm)
GC ≥98.5 %