1,10-fenanthrolinhydrokloridmonohydrat ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate
A16254.06 A16254.14 A16254.22
A16254.06EA 725 SEK
A16254.06 A16254.14 A16254.22
1,10-fenanthrolinhydrokloridmonohydrat ≥99%
1,10-fenanthrolinhydrokloridmonohydrat
Purity: 99% CAS No.: 18851-33-7 Molecular Formula: C12H8N2.HCl.H2O Formula Weight: 234.68 (216.66anhy) Odour: Odourless Melting Point: ca 225° dec. Beilstein Reference: 4007967 UN No.: 2811 MDL No.: MFCD00150061
Formel: C₁₂H₉ClN₂·1H₂O
MW: 234.68 g/mol
Smältpunkt: ∼ 225 °C (dec.)
Densitet: 1.385 g/cm³ (–100.15 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00150061
CAS nummer: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1 III