Melanin 99%, Sigma-Aldrich®

Leverantör: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

M2649-100MG M2649-1G
M2649-100MGEA 1000 SEK
M2649-100MG M2649-1G
Melanin 99%, Sigma-Aldrich®
Melanin

Melanin is a ubiquitous and prominent pigment responsible for color of skin, hair and eyes. Melanin is classified into eumelanin and pheomelanin. Eumelanin is black to dark brown pigment and is present in black hair and retina of the eye. Pheomelanin is yellow to reddish brown pigment present is red hair and red feathers. Melanins are high molecular weight pigments synthesised from phenolic or indolic precursors and is widely present in animals, plants and microorganisms.


  • Melanin from Sepia officinalis has been used as a natural pigment for Raman spectroscopy measurements
  • In competitive enzyme linked immunosorbent assay (ELISA) for the characterisation of phage antibodies
  • As an external standard for spectrophotometrically determining the melanin production in B16F10 cells

Formel: C₁₈H₁₀N₂O₄
Storage Temperature: Frys
MDL Number: MFCD00131581
CAS nummer: 8049-97-6

Order Now

Specifikation testresultat

Appearance Black Powder
Solubility Dark brown solution with a few insolubles at 10 mg plus 10 ml of 1% hydrogen peroxide, Ph 10
IR Spectrum Consistent with structure
Purity by thin layer chromatography Greater than 99%
Qc Acceptance Date Jun-03


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR