Tymolblått natriumsalt
Leverantör: Thermo Fisher Scientific
Synonyms: Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bis[5-methyl-2-(1-methylethyl)phenolate] S,S-dioxide, Sodium 4-(3-(4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-isopropyl-5-methylphenolate, Thymol blue water soluble, Thymol Blue sodium
B23007.09 B23007.18 B23007.30
B23007.09EA 660 SEK
B23007.09 B23007.18 B23007.30
Tymolblått natriumsalt
Tymolblått natriumsalt
CAS No.: 62625-21-2 Molecular Formula: C27H29NaO5S Formula Weight: 488.58 Melting Point: ca 284° dec. Beilstein Reference: 3861515 MDL No.: MFCD00151093
Formel: C₂₇H₂₉NaO₅S
MW: 488.58 g/mol
Smältpunkt: ∼ 284 °C (dec.)
Densitet: 1.19 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00151093
CAS nummer: 62625-21-2
EINECS: 263-650-6