Natriumstannat trihydrat ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Sodium tin(IV) oxide trihydrate, Disodium tin trioxide trihydrate
A18611.0E A18611.22 A18611.36
A18611.0EEA 9675 SEK
A18611.0E A18611.22 A18611.36
Natriumstannat trihydrat ≥98%
Natriumstannat trihydrat
Purity: 98% CAS No.: 12209-98-2 Molecular Formula: Na2SnO3.3H2O Formula Weight: 266.75 (212.67anhy) Odour: Odourless Merck Reference: 14,8677 MDL No.: MFCD00150189
Formel: Na₂O₃Sn·3H₂O
MW: 266.73 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00150189
CAS nummer: 12209-98-2
EINECS: 235-030-5