Devardas metallbad (Cu:Al:Zn 50:45:5 wt%), granulat
Leverantör: Thermo Scientific
Fara
Synonyms: Copper aluminium zinc alloy (50:45:5 w%), Devarda's metal
45526.22 45526.36
45526.22EA 1425 SEK
45526.22 45526.36
Devardas metallbad (Cu:Al:Zn 50:45:5 wt%), granulat
Devardas metallbad
CAS No.: 8049-11-4 Molecular Formula: Cu:Al:Zn; 50:45:5 wt% Density: 4.355 Merck Reference: 14,2941 UN No.: 3089 MDL No.: MFCD00198176
Kokpunkt: 906 °C (1013 hPa)
Smältpunkt: 490…560 °C
Densitet: 4,355
Storage Temperature: Omgivande
MDL Number: MFCD00198176
CAS nummer: 8049-11-4
UN: 3089
ADR: 4.1,II
Merck Index: 14,2941